Learn More
CENPJ Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL594-11517-100UL
This item is not returnable.
View return policy
Description
CENPJ, also named as CPAP, LAP and LIP1, belongs to the TCP10 family. It plays an important role in cell division and centrosome function by participating in centriole duplication. CENPJ inhibits microtubule nucleation from the centrosome.
Specifications
CENPJ | |
Polyclonal | |
CoraLite 594 | |
100 μL | |
55835 | |
Liquid |
Immunofluorescence | |
1000 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse | |
IgG |