missing translation for 'onlineSavingsMsg'
Learn More

Cellulose microcrystalline, extra pure, average particle size 90μm, Thermo Scientific Chemicals

Product Code. 11916991
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Glass bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-34-6

Specifications

Color White
pH 5 to 7
Physical Form Fine Crystalline Powder or Granular Powder
Linear Formula (C12H20O10)n
Solubility Information Solubility in water: insoluble.
Quantity 5 kg
Grade Extra Pure
Infrared Spectrum Authentic
Loss on Drying 1.5% max.
Packaging Plastic drum
Particle Size 250 Micron <1% min., 75 Micron >45% min., 32 Micron >70% min.
Residue after Ignition 0.05% max.
Chemical Name or Material Cellulose microcrystalline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'