missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Cell Culture Flasks with Plug Seal Cap, Tissue Culture Treated

Preferred vessel for long term cultivation or large-scale expansion of cells, providing protection against contamination. Eppendorf™ Cell Culture Flasks with Plug Seal Cap, Tissue Culture Treated are sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free and non-cytotoxic.

Specifications

Color Transparent
Material Polystyrene
For Use With (Application)
  • Expansion and culture of adherent and suspension cells on TC treated or on non-treated polystyrene
  • Performance of cell-based assays and microscopic analysis
Surface Coating Tissue Culture Treated
Temperature Range (Metric) -86°C to +60°C
View More Specs

Description

Description

  • Features ConvexAcces™ geometry for facilitated access to growth area and easier and safe cell treatment
  • Defined arrested position on plug-seal caps to prevent undesired closing
  • Anti-rolling cap with corrugation facilitates cultivation steps when cap needs to be set aside
  • 100% in-line control for leakage-free flasks and maximum safety
  • Direct surface identification on flask and cap
  • Available in three sizes: T–25, T–75 and T–175 with TC treated surfaces
Specifications

Specifications

Transparent
  • Expansion and culture of adherent and suspension cells on TC treated or on non-treated polystyrene
  • Performance of cell-based assays and microscopic analysis
-86°C to +60°C
SAL 10-6
Polystyrene
Tissue Culture Treated
Not Autoclavable
Videos
SDS
Documents

Documents

Product Certifications