missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDH5 Mouse anti-Human, Clone: 4F9B10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66804-1-IG-150UL
This item is not returnable.
View return policy
Description
VE-cadherin is a member of the cadherin superfamily that is located in a six-cadherin cluster in a region on the long arm of chromosome 16 and is involved in loss of heterozygosity events in breast and prostate cancer. VE-cadherin protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Functioning as a classic cadherin by imparting to cells the ability to adhere in a homophilic manner, VE-cadherin may play an important role in endothelial cell biology through control of the cohesion and organization of the intercellular junctions. An alternative splice variant has been described but the full length sequence of VE-cadherin has not been determined.Specifications
CDH5 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P33151 | |
CDH5 | |
CDH5 Fusion Protein Ag27501 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin) | |
4F9B10 | |
Unconjugated | |
CDH5 | |
7B4, 7B4 antigen, Cadherin 5, CD144, CDH5, FLJ17376, Vascular endothelial cadherin, VE cadherin | |
Mouse | |
Protein G | |
RUO | |
1003 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction