Learn More
CD8a Mouse anti-Human, FITC, Clone: OKT8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech FITC-65135-100TESTS
This item is not returnable.
View return policy
Description
The CD8a antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD8a. This antibody recognizes human antigen. The CD8a antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifications
CD8a | |
Monoclonal | |
5 μL (0.125 μg)/Test | |
Mouse | |
Primary | |
Human |
Flow Cytometry | |
OKT8 | |
FITC | |
100 Tests | |
925 | |
IgG2a |