Learn More
CD4 Rat anti-Mouse, PE, Clone: GK1.5, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech PE-65104-100UG
This item is not returnable.
View return policy
Description
The CD4 antibody from Proteintech is a rat monoclonal antibody generated with mouse ctl clone v4 of mouse CD4. This antibody recognizes mouse antigen. The CD4 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifications
CD4 | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
Primary | |
Mouse |
Flow Cytometry | |
GK1.5 | |
PE | |
100 μg | |
12504 | |
IgG2b |