Learn More
IBA Lifesciences CD4 Fab-TACS™ Agarose Column Starter Kit
Label-free isolation of CD4+ T cells
154.00 € - 265.50 €
Specifications
Buffer 10 x Buffer CI
View More Specs
Products 2
Product Code Brand Target Species Price Quantity & Availability  
Product Code Brand Target Species Price Quantity & Availability  
17227740
View Documents
IBA Lifesciences
6-3202-002
Human This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17267740
View Documents
IBA Lifesciences
6-3301-002
Mouse This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Fab-TACS™ - Traceless Affinity Cell Selection - is a positive cell isolation approach based on our Strep-tag™ technology
  • CD4 is a co-receptor that associates with the T cell receptor and interacts with MHC class II molecules
  • Strep-Tactin™ TACS Agarose columns are filled with Strep-Tactin™ coated agarose macrobeads developed for cell isolation. Fab fragments fused to a Twin-Strep-tag™ (Fab-Streps) bind to the matrix.
  • Target cells adhere to the matrix based on the exclusive binding of the Fab-Strep to a surface marker. Non-target cells are efficiently washed away.
  • In a final step, the addition of biotin causes the elution of the target cells and the Fab-Streps. After elution, the Fab-Streps self-dissociate from the cell surfaces due to their reduced affinity
  • The label-free authentic target cells are now ready for further downstream applications
Specifications
Specifications
10 x Buffer CI