Learn More
IBA Lifesciences CD31 Isolation Kit for FABian™
For automated positive cell separation. The Isolation Kit for FABian™ provides label-free target cells without the use of high affinity antibodies and magnetic beads
Brand: IBA Lifesciences 6-6016-001
This item is not returnable.
View return policy
Description
Specifications
Non-magnetic Cell Isolation Kit | |
Human Whole Blood or Buffy Coat | |
9 mL Whole Blood |
2°C to 8°C | |
Isolation of label-free cells | |
1 Kit, Human |