Learn More
CD3 Mouse anti-Human, CoraLite 647, Clone: Hit3a, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL647-65112-100TESTS
This item is not returnable.
View return policy
Description
The CD3 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD3. This antibody recognizes human antigen. The CD3 antibody has been validated for the following applications: FC analysis. This antibody is coralite™647.
Specifications
CD3 | |
Monoclonal | |
5 μL/Test | |
Mouse | |
Primary | |
Human |
Flow Cytometry | |
Hit3a | |
CoraLite 647 | |
100 Tests | |
916 | |
IgG2a |