missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD3 Fab-TACS™ Gravity Kit
IBA Lifesciences™ CD3 Fab-TACS™ Gravity Kit is for reversible cell isolation delivering label-free target cells.
361.00 € - 829.00 €
Specifications
Product Type | Non-magnetic Cell Isolation Kit |
---|---|
Content And Storage | 2°C to 8°C |
Donor Source | Human Whole Blood |
For Use With (Application) | Isolation of label-free cells |
Description
- The kit allows the selection of CD3+ T cells directly from whole blood.
- CD3 Fab-Strep, human
- Buffer CI (10x)
- Biotin stock solution
- Fab-TACS™ Gravity Adapter
- Fab-TACS™ Flow Restrictor
- The Fab-TACS™ Gravity column is filled with a Strep-Tactin™ coated matrix made of cell-grade agarose.
- Strep-tagged Fab fragments (Fab-Streps) specifically bind to the matrix.
- Subsequently, whole blood or other blood preparations pass through the column.
- Target cells adhere to the matrix based on the exclusive binding of the Fab-Strep to the target cell.
- Non-target cells are efficiently washed away.
- In a final step, the addition of biotin causes the elution of the target cells and the Fab-Streps.
- After elution, the Fab-Streps self-dissociate from the cell surfaces.
- The label-free authentic target cells are now ready for further downstream applications.
Specifications
Non-magnetic Cell Isolation Kit | |
2°C to 8°C | |
Human Whole Blood | |
Isolation of label-free cells |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title