missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD28 Isolation Kit for FABian™
IBA Lifesciences™ CD28 Isolation Kit for FABian™ is for reversible cell isolation delivering label-free target cells.
90.41 € - 718.00 €
Specifications
Product Type | Non-magnetic Cell Isolation Kit |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Application) | Isolation of label-free cells |
Description
- The kit is used for the selection of CD28+ T cells from whole blood or buffy coat in conjunction with the FABian™ system.
- It includes CD28 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
- Each kit is sufficient for one FABian™ run to isolate cells from up to 9mL whole blood or up to 6.25mL buffy coat.
Specifications
Non-magnetic Cell Isolation Kit | |
2°C to 8°C | |
Isolation of label-free cells |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title