missing translation for 'onlineSavingsMsg'
Learn More

CD206 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen CD206
Concentration 0.25 mg/mL
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16818584
View Documents Promotion Details
Proteintech
18704-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16808584
View Documents Promotion Details
Proteintech
18704-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody recognizes the intracellular carboxy-terminal part of CD206 and MRC1L1.

CD206 (macrophage mannose receptor, MMR), also known as mannose receptor C1 (MRC1), is a type I transmembrane glycoprotein serving as pattern recognition receptor for carbohydrate groups on the surface of bacteria, fungi and other pathogens. Expressed mainly on tissue macrophages and dendritic cells, CD206 mediates endocytosis of these pathogens and presentation of their antigens to the adaptive immune system. CD206 can also be detected in a soluble form in human plasma and is elevated in patients with acute sepsis.
Specifications

Specifications

CD206
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Mouse, Human, Rat
P22897, Q61830
17533, 291327, 4360
Peptide
Primary
-20°C
MRC1
0.25 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CD206, CLEC13D, Macrophage mannose receptor 1, Mannose receptor, mannose receptor, C type 1, MMR, MRC1
Mrc1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications