missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD147 Mouse anti-Human, Porcine, Clone: 1G12B5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66443-1-IG-150UL
This item is not returnable.
View return policy
Description
CD147 (basigin, neurothelin, OX-47, 5A11, CE9, M6, EMMPRIN, extracellular matrix metalloproteinase inducer, TCSF, tumour cell-derived collagenase-stimulatory factor) is a 50-60 kDa glycoprotein with multiple glycosylated forms. CD147 is a plasma membrane protein that is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. CD147 is also a member of the immunoglobulin superfamily. The highest level of CD147 expression is on metabolically active cells, such as lymphoblasts, inflammatory cells, brown adipocytes and maligt tumor cells. CD147 has multiple functions, including facilitating of cell surface expression of monocarboxylate transporter proteins and extracellular matrix metalloproteinases, regulation of integrin functions, it plays roles in cell development and activation, fetal development or retinal function.Specifications
CD147 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35613 | |
BSG | |
CD147 Fusion Protein Ag2619 | |
150 μL | |
Primary | |
Human, Porcine | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1G12B5 | |
Unconjugated | |
BSG | |
5F7, Basigin, basigin (Ok blood group), BSG, CD147, Collagenase stimulatory factor, EMMPRIN, HAb18G, M6, OK, OK blood group antigen, TCSF | |
Mouse | |
Protein G | |
RUO | |
100141312, 682 | |
-20°C | |
Liquid |