missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences CD14 Isolation Kit for FABian™

IBA Lifesciences™ CD14 Isolation Kit for FABian™ is for reversible cell isolation delivering label-free target cells.

90.41 € - 718.00 €

Specifications

Product Type Cell Isolation Kit
Content And Storage 2°C to 8°C
Donor Source Human Whole Blood
For Use With (Application) Isolation of label-free cells
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15829784
View Documents
IBA Lifesciences
6-6017-001
1 Kit, Human This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15839784
View Documents
IBA Lifesciences
6-6017-010
10 Kits, Human This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • The kit is used for the selection of CD14+ cells from buffy coat or whole blood in conjunction with the FABian™ system.
  • It includes CD14 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
  • Each kit is sufficient for one FABian™ run to isolate cells from 4.25mL buffy coat.
  • Alternatively 9mL whole blood may be used.
Specifications

Specifications

Cell Isolation Kit
2°C to 8°C
Human Whole Blood
Isolation of label-free cells
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.