missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11d/Integrin alpha D Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 410.00 €
Specifications
Antigen | CD11d/Integrin alpha D |
---|---|
Concentration | 0.16 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16816645
|
Proteintech
55162-1-AP-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16806645
|
Proteintech
55162-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is specific to CD11d.
Integrin alpha-D/beta-2 is a receptor for ICAM3 and VCAM1. May play a role in the atherosclerotic process such as clearing lipoproteins from plaques and in phagocytosis of blood-borne pathogens, particulate matter, and senescent erythrocytes from the blood.Specifications
CD11d/Integrin alpha D | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q13349 | |
3681 | |
Peptide | |
Primary | |
-20°C | |
ITGAD |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ADB2, CD11d, Integrin alpha D, integrin, alpha D, ITGAD, Leukointegrin alpha D | |
ITGAD | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title