Learn More
CD11b Mouse anti-Human, PE, Clone: ICRF44, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech PE-65116-100TESTS
Product Code. 16801716
  • 80.00 € / 100 tests
Explore more special offers
Description
Description
The CD11b antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD11b. This antibody recognizes human antigen. The CD11b antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifications
Specifications
CD11b
Monoclonal
5 μL (1 μg)/Test
Mouse
Primary
Human
Flow Cytometry
ICRF44
PE
100 Tests
3684
IgG1