missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Casein, Vitamin Tested, 30 Mesh
Used as cheese, food additive and binder for safety matches.
344.00 € - 4835.00 €
Specifications
Product Type | Casein |
---|---|
Color | Yellow |
Format | Powder |
For Use With (Application) | Used as cheese, food additive and binder for safety matches |
Storage Requirements | Store at room temperature (15°C to 30°C) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11455722
|
MP Biomedicals™
0290452005 |
5 lb. |
344.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11465722
|
MP Biomedicals™
0290452025 |
25 lb. |
1545.00 €
25lb |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11432490
|
MP Biomedicals™
0290452080 |
100 lb. |
4835.00 €
100lb |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Phosphoprotein
- Extracted from bovine milk
- Used as food source
- CAS: 9000-71-9
Specifications
Casein | |
Powder | |
Store at room temperature (15°C to 30°C) |
Yellow | |
Used as cheese, food additive and binder for safety matches |