missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Cardboard Cryogenic Vial Box Partition, 7 x 7
GREENER_CHOICE

Creates a 49-well interior configuration (well diameter 17.5 mm) for cryogenic storage of 1.5 / 2.0 mL vials in mechanical freezers or vapor phase LN2.

Brand:  Heathrow Scientific™ HS2860G

Product Code. 15740039

  • 32.20 € / Pack of 12
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Store in mechanical freezers or vapor phase LN2, -196°C
  • Color coded boxes allow for easy identification and sorting purposes
  • Bottom vent holes allow quicker circulation and removal of cold air, vapor or LN2
  • All box footprints are 13.3 x 13.3 cm, select from two different heights
  • Must use tubes capable of being stored at the required temperature of the user
  • Not recommended for autoclaving.
  • Partitions and Boxes/Lids must be ordered separately
Specifications

Specifications

White
Square
Cryogenic Storage
49 (17.5 mm Diameter Vials)
7 x 7
13.3 cm
17.5 mm
Cardboard
Vial Box Partition
Cardboard Cryogenic Storage Box and Lid
12 Pack
13.3 x 13.3 cm
13.3 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.