missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonate solution, 0.35M Carb./0.10M Bicarb., for Ion Chromatography, 100mg/L, H2O matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
74.00 € - 135.00 €
Specifications
Solution Contains | 0.35M Carbonate/0.10M Bicarbonate |
---|---|
Physical Form | Liquid |
Concentration | 100 mg/L |
Type | Single-component Standard Solution |
Solvent or Matrix | H2O |
Chemical Name or Material | Carbonate solution |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12922245
![]() |
Fisher Chemical
A 106 100 |
100 mL | Bottle |
74.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12933115
![]() |
Fisher Chemical
A 106 500 |
500 mL | Bottle |
135.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
0.35M Carbonate/0.10M Bicarbonate | |
100 mg/L | |
H2O |
Liquid | |
Single-component Standard Solution | |
Carbonate solution |
Safety and Handling
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Carbonate solution, 0.35M Carb./0.10M Bicarb., for Ion Chromatography, 100mg/L, H2O matrix, Fisher chemical™