missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Carbonate, 0.5M buffer soln., pH 9.0
73.67 € - 114.57 €
Specifications
Concentration | 0.5 M buffer soln. |
---|---|
pH Range | 9.0 |
Type | Carbonate buffer soln. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15426059
|
Thermo Scientific Chemicals
J63899.AK |
250 mL |
73.67 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15436059
|
Thermo Scientific Chemicals
J63899.AP |
500 mL |
114.57 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
0.5 M buffer soln. | |
9.0 | |
Carbonate buffer soln. |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals Carbonate, 0.5M buffer soln., pH 9.0