Learn More
Carbon tetrabromide, 98%, Thermo Scientific Chemicals
40.60 € - 212.06 €
Chemical Identifiers
CAS | 558-13-4 |
---|---|
Molecular Formula | CBr4 |
Molecular Weight (g/mol) | 331.64 |
MDL Number | MFCD00000117 |
InChI Key | HJUGFYREWKUQJT-UHFFFAOYSA-N |
Synonym | carbon tetrabromide, methane, tetrabromo, carbon bromide, methane tetrabromide, bromid uhlicity, carbontetrabromide, methane, tetrabromide, tetrabrommethan, bromid uhlicity czech |
PubChem CID | 11205 |
ChEBI | CHEBI:47875 |
IUPAC Name | tetrabromomethane |
SMILES | C(Br)(Br)(Br)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10013590
|
Thermo Scientific Acros
325150050 |
5 g | Glass Bottle |
40.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10431121
|
Thermo Scientific Acros
325151000 |
100 g | Plastic bottle |
63.92 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10431311
|
Thermo Scientific Acros
325155000 |
500 g | Plastic bottle |
212.06 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
558-13-4 | |
331.64 | |
HJUGFYREWKUQJT-UHFFFAOYSA-N | |
11205 | |
tetrabromomethane |
CBr4 | |
MFCD00000117 | |
carbon tetrabromide, methane, tetrabromo, carbon bromide, methane tetrabromide, bromid uhlicity, carbontetrabromide, methane, tetrabromide, tetrabrommethan, bromid uhlicity czech | |
CHEBI:47875 | |
C(Br)(Br)(Br)Br |
Specifications
558-13-4 | |
White | |
97.5% min. (On organic basis) (GC) | |
CBr4 | |
01, 68 | |
carbon tetrabromide, methane, tetrabromo, carbon bromide, methane tetrabromide, bromid uhlicity, carbontetrabromide, methane, tetrabromide, tetrabrommethan, bromid uhlicity czech | |
HJUGFYREWKUQJT-UHFFFAOYSA-N | |
tetrabromomethane | |
11205 | |
331.64 | |
Crystalline Solid |
88°C to 93°C | |
190°C | |
CBr4 | |
MFCD00000117 | |
15,352; 16,366 | |
Solubility in water: insoluble | |
C(Br)(Br)(Br)Br | |
331.64 | |
CHEBI:47875 | |
98% | |
Carbon tetrabromide |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medi
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-189-6
- Carbon tetrabromide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only