missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 7440-44-0 |
---|---|
Molecular Formula | C |
Molecular Weight (g/mol) | 12 |
MDL Number | MFCD00133992 |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
Synonym | graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve |
PubChem CID | 5462310 |
ChEBI | CHEBI:27594 |
IUPAC Name | carbon |
SMILES | [C] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aidChemical Identifiers
7440-44-0 | |
12 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
5462310 | |
carbon |
C | |
MFCD00133992 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
CHEBI:27594 | |
[C] |
Specifications
7440-44-0 | |
Black | |
15% max. | |
MFCD00133992 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
12 | |
Activated charcoal,NORIT* CN 1, NORIT CN 1 |
3652.0°C | |
5.5 to 8 | |
C | |
15, 1807 | |
Solubility in water: insoluble. Other solubilities: insoluble in organic solvents | |
[C] | |
12 | |
CHEBI:27594 | |
Powder |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 231-153-3
missing translation for 'rtecsNumber' : FF5250100
missing translation for 'tsca' : TSCA
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Activated charcoal
Signal Word
- Warning
Hazard Category
- Self-heating substance Category 2
Hazard Statement
- H252-Self-heating in large quantities; may catch fire.
Precautionary Statement
- P235+P410-Keep cool. Protect from sunlight.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title