Learn More
CAPZA1 Mouse anti-Human, CoraLite 488, Clone: 10H1F9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66066-100UL
Product Code. 16810716
  • 450.00 € / 100µL
Explore more special offers
Description
Description
The CAPZA1 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human CAPZA1. This antibody recognizes human antigen. The CAPZA1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
CAPZA1
Monoclonal
CoraLite 488
100 μL
829
IgG1
Immunofluorescence
10H1F9