Learn More
Erlab™ Captair™ Smart 633 Ductless Fume Hood
Designed with advanced carbon filtration to minimize chemical exposure.
Specifications
Height (Metric) 111 cm
View More Specs
Description
Description
  • Sensors to detect filter breakthrough of solvents, acids or formaldehyde
  • Safety back up filter in case of main filter saturation
  • Continuous monitoring of air face velocity
Erlab Safety Program
  • Application analysis and validation, usage certification, filter change reminders
  • Connect with eGuard™ for real-time status, notifications and alerts
  • Smart-Light feature for lighting up the hood
Flexibility
  • Configurable filtration column
  • No ductwork needed
Environment
  • No dangerous chemicals are released into atmosphere
  • Low energy consumption
Specifications
Specifications
111 cm