missing translation for 'onlineSavingsMsg'
Learn More
Learn More
behr Labor-Technik™ HDPE, Canister with Screw Cap and UN Approval
Use these canisters for a variety of applications. behr Labor-Technik™ Canisters are ideal for the transport and storage of hazardous substances. Available in a variety of sizes.
Specifications
Certifications/Compliance | UN-certified |
---|---|
Closure Type | Screw Cap |
Color | White |
Material | HDPE |
Product Type | Canister |
Description
- White High-Density Polyethylene canisters
- UN-certified
- Screw-on cap
- Suitable for hazardous goods
Specifications
UN-certified | |
White | |
Canister |
Screw Cap | |
HDPE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title