Learn More
Thermo Scientific Chemicals Calpain Inhibitor XI
81.81 € - 365.00 €
Chemical Identifiers
CAS | 145731-49-3 |
---|---|
Molecular Formula | C26H40N4O6 |
Synonym | Z-Leu-α-aminobutyric acid-CONH(CH2)3-morpholine; Z-L-Abu-CONH(CH2)3-morpholine |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15417179
|
Thermo Scientific Chemicals
J65991.M^ |
1 mg |
81.81 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15407179
|
Thermo Scientific Chemicals
J65991.#0 |
5 mg |
365.00 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A cell-permeable dipeptidyl --ketoamide that acts as a potent, highly selective, reversible, and active site inhibitor of calpain-1 and -2
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
145731-49-3 | |
C26H40N4O6 | |
Z-Leu-α-aminobutyric acid-CONH(CH2)3-morpholine; Z-L-Abu-CONH(CH2)3-morpholine |
Specifications
Off-white | |
A cell-permeable dipeptidyl →-ketoamide that acts as a potent, highly selective, reversible, and active site inhibitor of calpain-1 and -2 | |
C26H40N4O6 | |
Z-Leu-α-aminobutyric acid-CONH(CH2)3-morpholine; Z-L-Abu-CONH(CH2)3-morpholine | |
Solid | |
CCC(NC(=O)C(CC(C)C)NC(=O)OCC1=CC=CC=C1)C(=O)C(=O)NCCCN1CCOCC1 | |
504.63 |
2°C to 8°C | |
145731-49-3 | |
504.60 | |
Light Sensitive | |
TZVQRMYLYQNBOA-UHFFFAOYNA-N | |
benzyl N-{3-methyl-1-[(1-{[3-(morpholin-4-yl)propyl]carbamoyl}-1-oxobutan-2-yl)carbamoyl]butyl}carbamate |
Safety and Handling
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.