Learn More
CALN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11477-1-AP-20UL
 View more versions of this product
Product Code. 16858583
  • 145.00 € / 20µL
Explore more special offers
Description
Description
Calcium binding proteins (CaBP) play a crucial role in the calcium-mediated cellular signal transduction pathway in the central nervous system. The CaBP family shares much similarity with CaM I (calmodulin), and it has been shown that CaBP proteins can substitute functionally for, and possibly augment the function of, CaM I. CaBP8, contains two EF-hand domains for calcium binding and shares 70% homology with CaBP7 and 50% homology with CaM I. It negatively regulates Golgi-to-plasma membrane trafficking by interacting with PI4KB and inhibiting its activity. Both CaBP8 and CaBP7 possess a targeting mechanism that is unique amongst the CaBPs that may contribute to differential functional Ca2+-sensing by these family members.
Specifications
Specifications
CALN1
Polyclonal
Unconjugated
CALN1
CaBP8, Calcium binding protein 8, CALN1, calneuron 1, Calneuron I
Rabbit
Antigen Affinity Chromatography
RUO
140904, 363909, 83698
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.53 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q06BI3, Q9BXU9, Q9JJG7
Caln1
CALN1 Fusion Protein Ag2043
20 μL
Primary
Rat, Human, Mouse