Learn More
BINDER™ Calibration Certificate, Temperature
Calibration Certificate for temperature, measurement in center of chamber at specified temperature.
Brand:  BINDER™ 8012-1609
 View more versions of this product
Product Code. 15923012
  • 1915.00 € / Each
Explore more special offers
Description
Description
  • Measurement at specified temperature
  • Including certificate
  • Various points of measurement
Specifications
Specifications
BINDER Chambers (Models MKF 56, MKF 115, MKF 240, MKF 720)
27 Measurement Points
Temperature Measurement
Calibration certificate, temperature - temperature measurement incl. certificate and 27 measuring points (at specified temperature)
Calibration Certificate (Temperature)