missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 4.01 and 7.01 Calibration Buffer Sachets, Hanna Instruments™
Click to view available options
Quantity:
20 mL
Description
- Set of premium quality pH 4.01 and 7.01 calibration buffer solutions that is NIST traceable and supplied with a Certificate of Analysis
- Supplied with Certificate of Analysis
- Accuracy of +/- 0.01 pH @ 25°C
- Temperature chart of the actual pH value at various temperatures printed on each bottle
- Set of premium quality pH 4.01 and 7.01 calibration buffers produced in accordance with ISO 3696/BS3978 standards
- Uses high purity salts, deionized water, certified weight-checked balances, and Class A glassware
- Temperature controlled environment monitored with certified thermometers.
- Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
Specifications
Specifications
pH | 7.01, 4.01 |
Quantity | 20 mL |
Solution Type | Buffer |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'