missing translation for 'onlineSavingsMsg'
Learn More

Calcium tungsten oxide, 98%, Thermo Scientific Chemicals

Product Code. 11302138
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Calcium tungsten oxide is used as a luminophore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-75-2
MDL Number MFCD00010913
Synonym Calcium tungstate

Specifications

Melting Point 1620°C
Physical Form Powder
Quantity 100 g
Merck Index 14,1712
Solubility Information Insoluble in H2O; Decomposes by hot HCl,HNO3
Formula Weight 287.93
Percent Purity 98%
Chemical Name or Material Calcium tungsten oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'