Learn More
Calcium trifluoromethanesulfonate, 99% min, Thermo Scientific Chemicals
110.00 € - 716.00 €
Chemical Identifiers
CAS | 55120-75-7 |
---|---|
Molecular Formula | C2CaF6O6S2 |
Molecular Weight (g/mol) | 338.204 |
MDL Number | MFCD00143911 |
InChI Key | PUQLFUHLKNBKQQ-UHFFFAOYSA-L |
Synonym | calcium trifluoromethanesulfonate, calcium triflate, methanesulfonic acid, trifluoro-, calcium salt, calcium trifluoromethansulphonate, methanesulfonic acid, 1,1,1-trifluoro-, calcium salt 2:1, calcium ditriflate, calcium trifluoromethyl sulfonate, calcium ii triflate, calcium 2+ ditriflate |
PubChem CID | 2734307 |
IUPAC Name | calcium;trifluoromethanesulfonate |
SMILES | C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Ca+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11379277
|
Thermo Scientific Alfa Aesar
040134.09 |
10 g |
110.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11369277
|
Thermo Scientific Alfa Aesar
040134.18 |
50 g |
365.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11359277
|
Thermo Scientific Alfa Aesar
040134.22 |
100 g |
716.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium trifluoromethanesulfonate is used for aminolysis of epoxides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
55120-75-7 | |
338.204 | |
PUQLFUHLKNBKQQ-UHFFFAOYSA-L | |
2734307 | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Ca+2] |
C2CaF6O6S2 | |
MFCD00143911 | |
calcium trifluoromethanesulfonate, calcium triflate, methanesulfonic acid, trifluoro-, calcium salt, calcium trifluoromethansulphonate, methanesulfonic acid, 1,1,1-trifluoro-, calcium salt 2:1, calcium ditriflate, calcium trifluoromethyl sulfonate, calcium ii triflate, calcium 2+ ditriflate | |
calcium;trifluoromethanesulfonate |
Specifications
(decomposition) | |
White | |
C2CaF6O6S2 | |
MFCD00143911 | |
Soluble in water. | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Ca+2] | |
338.204 | |
338.21 | |
Calcium trifluoromethanesulfonate |
55120-75-7 | |
Powder | |
Ca(OSO2CF3)2 | |
calcium trifluoromethanesulfonate, calcium triflate, methanesulfonic acid, trifluoro-, calcium salt, calcium trifluoromethansulphonate, methanesulfonic acid, 1,1,1-trifluoro-, calcium salt 2:1, calcium ditriflate, calcium trifluoromethyl sulfonate, calcium ii triflate, calcium 2+ ditriflate | |
PUQLFUHLKNBKQQ-UHFFFAOYSA-L | |
calcium;trifluoromethanesulfonate | |
2734307 | |
≤99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 232-188-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only