missing translation for 'onlineSavingsMsg'
Learn More

Calcium titanium oxide, 99+% (metals basis), Thermo Scientific Chemicals

162.81 € - 538.68 €

Chemical Identifiers

CAS 12049-50-2
Molecular Formula CaO3Ti
Molecular Weight (g/mol) 135.94
MDL Number MFCD00015981
InChI Key WEUCVIBPSSMHJG-UHFFFAOYSA-N
Synonym calcium titanate, titaniumcalciumoxide, calciumtitaniumtrioxide, ca.o3ti, titanatecalcium 1:1, calcium dioxido oxo titanium, calcium oxotitaniumbis olate, calcium 2+ oxotitaniumbis olate, titanate tio32-,calcium 1:1, calcium titanate, nanopowder, <100 nm particle size bet trace metals basis
PubChem CID 16212381
IUPAC Name calcium;dioxido(oxo)titanium
SMILES [O--].[O--].[O--].[Ca++].[Ti+4]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11311199
View Documents
Thermo Scientific Alfa Aesar
011397.36
500 g
162.81 €
500g
Please to purchase this item. Need a web account? Register with us today!
11301199
View Documents
Thermo Scientific Alfa Aesar
011397.A3
2 kg
538.68 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Calcium titanium oxide is used to prepare the ferrotitanium alloys and its structure is used to study the other compounds such as barium titanate.It is used to make the pure substances at low temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12049-50-2
135.94
WEUCVIBPSSMHJG-UHFFFAOYSA-N
16212381
[O--].[O--].[O--].[Ca++].[Ti+4]
CaO3Ti
MFCD00015981
calcium titanate, titaniumcalciumoxide, calciumtitaniumtrioxide, ca.o3ti, titanatecalcium 1:1, calcium dioxido oxo titanium, calcium oxotitaniumbis olate, calcium 2+ oxotitaniumbis olate, titanate tio32-,calcium 1:1, calcium titanate, nanopowder, <100 nm particle size bet trace metals basis
calcium;dioxido(oxo)titanium

Specifications

1975°C
Beige
>99% (Metals basis)
MFCD00015981
Insoluble in water. Soluble in oirganic and inorganic solution.
[O--].[O--].[O--].[Ca++].[Ti+4]
135.94
135.98
4.10 g/cm3
12049-50-2
Powder
CaO3Ti
calcium titanate, titaniumcalciumoxide, calciumtitaniumtrioxide, ca.o3ti, titanatecalcium 1:1, calcium dioxido oxo titanium, calcium oxotitaniumbis olate, calcium 2+ oxotitaniumbis olate, titanate tio32-,calcium 1:1, calcium titanate, nanopowder, <100 nm particle size bet trace metals basis
WEUCVIBPSSMHJG-UHFFFAOYSA-N
calcium;dioxido(oxo)titanium
16212381
Odorless
Calcium titanium oxide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 234-988-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only