Learn More
Calcium thioglycolate trihydrate, 99%, Thermo Scientific Chemicals
28.14 € - 102.51 €
Chemical Identifiers
CAS | 65208-41-5 |
---|---|
Molecular Formula | C2H10CaO5S |
Molecular Weight (g/mol) | 186.24 |
MDL Number | MFCD00150707 |
InChI Key | GDECCIJZCWLFSB-UHFFFAOYSA-N |
Synonym | calciumthioglycolatetrihydrate |
PubChem CID | 131881455 |
SMILES | O.O.O.[Ca].OC(=O)CS |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10743023
|
Thermo Scientific Acros
367031000 |
100 g | Plastic bottle |
28.14 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10018881
|
Thermo Scientific Acros
367035000 |
500 g | Plastic bottle |
102.51 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
65208-41-5 | |
186.24 | |
GDECCIJZCWLFSB-UHFFFAOYSA-N | |
131881455 |
C2H10CaO5S | |
MFCD00150707 | |
calciumthioglycolatetrihydrate | |
O.O.O.[Ca].OC(=O)CS |
Specifications
65208-41-5 | |
White | |
98.5% min. (Iodometry) | |
MFCD00150707 | |
calciumthioglycolatetrihydrate | |
GDECCIJZCWLFSB-UHFFFAOYSA-N | |
calcium;2-sulfanylacetic acid;trihydrate | |
131881455 | |
99% | |
Calcium thioglycolate trihydrate |
270°C to 280°C | |
Authentic | |
C2H10CaO5S | |
15, 1713 | |
Solubility in water: 70g/L (25°C). Other solubilities: very slightly soluble in alcohol,chloroform,practical insoluble in ether,petroleum ether,,benzene | |
O.O.O.[Ca].OC(=O)CS | |
186.24 | |
184.23 | |
Powder |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice
GHS Signal Word: Warning
- Calcium thioglycolate trihydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only