Learn More
Calcium sulfide, 99.9% (metals basis excluding Sr), Sr typically 500 ppm, Thermo Scientific Chemicals
67.60 € - 272.00 €
Chemical Identifiers
CAS | 20548-54-3 |
---|---|
Molecular Formula | CaS |
Molecular Weight (g/mol) | 72.14 |
MDL Number | MFCD00015990 |
InChI Key | AGVJBLHVMNHENQ-UHFFFAOYSA-N |
PubChem CID | 30182 |
IUPAC Name | calcium sulfanediide |
SMILES | [S--].[Ca++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11322218
|
Thermo Scientific Alfa Aesar
013121.06 |
5 g |
67.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11302218
|
Thermo Scientific Alfa Aesar
013121.14 |
25 g |
272.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as luminous paint, depilatory, lubricant additive, ore dressing and flotation agent and as an arsenic-free hydrogen sulfide. It is also used as a depilatory in the tanning industry and in cosmetics, in homeopathy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20548-54-3 | |
72.14 | |
AGVJBLHVMNHENQ-UHFFFAOYSA-N | |
calcium sulfanediide |
CaS | |
MFCD00015990 | |
30182 | |
[S--].[Ca++] |
Specifications
(decomposition) | |
Powder | |
MFCD00015990 | |
14,1707 | |
AGVJBLHVMNHENQ-UHFFFAOYSA-N | |
calcium sulfanediide | |
30182 | |
99.9% | |
Moisture sensitive | |
(metals basis excluding Sr), Sr typically 500ppm |
20548-54-3 | |
CaS | |
UN3134 | |
Slightly soluble in water. Insoluble in alcohol | |
[S--].[Ca++] | |
72.14 | |
72.14 | |
Stench | |
2.5 g/mL | |
Calcium sulfide |
Safety and Handling
GHS H Statement
H261-H315-H319-H335
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335-H500
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II
missing translation for 'einecsNumber' : 243-873-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only