missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium silicide, tech. Ca ≈30%, may contain up to 5% Fe, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
100 g
500 g
Description
It is an important raw material used in Pharmaceutical intermediates. It is an ideal compound of deoxidizer and desulfurizer which can be used in production of many types of stainless steel. It is also used as inoculants and additive in cast iron.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | ∼1000°C |
Color | Gray |
Physical Form | Powder |
Quantity | 10 g |
UN Number | UN1405 |
Solubility Information | Insoluble in cold water, decomposes in hot water |
Formula Weight | 96.25 |
Grade | Technical |
Concentration or Composition (by Analyte or Components) | Ca ≈30%, may contain up to 5% Fe |
Sensitivity | Moisture Sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'