Learn More
Calcium silicate, meta, 99% (metals basis), Thermo Scientific™
36.00 € - 417.00 €
Chemical Identifiers
CAS | 10101-39-0 |
---|---|
Molecular Formula | CaO3Si |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00015979 |
InChI Key | OYACROKNLOSFPA-UHFFFAOYSA-N |
Synonym | calcium metasilicate, wollastonite, aedelforsite, casiflux, cab-o-lite, gillebachite, wollastokup, calcium silicon oxide, florite, fuwalip |
PubChem CID | 26370 |
IUPAC Name | calcium;dioxido(oxo)silane |
SMILES | [O-][Si](=O)[O-].[Ca+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11308747
|
Thermo Scientific Alfa Aesar
035843.14 |
25 g | N/A | N/A | N/A | |||||
15161995
|
Thermo Scientific Alfa Aesar
035843.22 |
100 g | N/A | N/A | N/A | |||||
11398737
|
Thermo Scientific Alfa Aesar
035843.36 |
500 g | N/A | N/A | N/A | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
10101-39-0 | |
116.16 | |
OYACROKNLOSFPA-UHFFFAOYSA-N | |
26370 | |
[O-][Si](=O)[O-].[Ca+2] |
CaO3Si | |
MFCD00015979 | |
calcium metasilicate, wollastonite, aedelforsite, casiflux, cab-o-lite, gillebachite, wollastokup, calcium silicon oxide, florite, fuwalip | |
calcium;dioxido(oxo)silane |
Specifications
1540°C | |
White | |
99% | |
MFCD00015979 | |
Insoluble in water. | |
[O-][Si](=O)[O-].[Ca+2] | |
116.16 | |
116.16 | |
White powder | |
Calcium silicate, meta |
10101-39-0 | |
Powder | |
CaO3Si | |
calcium metasilicate, wollastonite, aedelforsite, casiflux, cab-o-lite, gillebachite, wollastokup, calcium silicon oxide, florite, fuwalip | |
OYACROKNLOSFPA-UHFFFAOYSA-N | |
calcium;dioxido(oxo)silane | |
26370 | |
Odorless | |
2.9 g/cm3 at 20°C |
Safety and Handling
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H335
missing translation for 'einecsNumber' : 233-250-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.