missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Nitrate Tetrahydrate, Honeywell Fluka™
Puriss. p.a., ACS Reagent, 99-103%
21.03 € - 322.11 €
Chemical Identifiers
CAS | 13477-34-4 |
---|---|
Molecular Formula | CaH8N2O10 |
Molecular Weight (g/mol) | 236.146 |
MDL Number | MFCD00149604 |
InChI Key | ICSSIKVYVJQJND-UHFFFAOYSA-N |
Synonym | calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate |
PubChem CID | 16211656 |
ChEBI | CHEBI:86159 |
IUPAC Name | calcium;dinitrate;tetrahydrate |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15673290
|
Honeywell-Fluka
31218-250G |
250 g | N/A | ||||||
15693290
|
Honeywell-Fluka
31218-500G |
500 g |
61.18 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15663290
|
Honeywell-Fluka
31218-1KG |
1 kg |
99.12 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15603300
|
Honeywell-Fluka
31218-5KG |
5 kg |
322.11 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
13477-34-4 | |
236.146 | |
ICSSIKVYVJQJND-UHFFFAOYSA-N | |
16211656 | |
calcium;dinitrate;tetrahydrate |
CaH8N2O10 | |
MFCD00149604 | |
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
CHEBI:86159 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] |
Specifications
13477-34-4 | |
Ca(NO3)2 · 4H2O | |
NONH for all modes of transport | |
ICSSIKVYVJQJND-UHFFFAOYSA-N | |
calcium;dinitrate;tetrahydrate | |
16211656 | |
236.15g/mol | |
ACS/puriss. p.a. |
CaH8N2O10 | |
MFCD00149604 | |
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] | |
236.146 | |
CHEBI:86159 | |
99 to 103% | |
Calcium Nitrate Tetrahydrate |
Safety and Handling
P280-P305 + P351 + P338
H302-H318
missing translation for 'einecsNumber' : 233-332-1
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title