missing translation for 'onlineSavingsMsg'
Learn More

Calcium Nitrate Tetrahydrate, Certified AR for Analysis, Fisher Chemical™

54.10 € - 99.50 €

Chemical Identifiers

CAS 13477-34-4
Molecular Formula CaH10N2O10
Molecular Weight (g/mol) 238.16
MDL Number 149604
InChI Key DKCJOKYHYIAZQH-UHFFFAOYSA-N
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.O.O.O.[Ca++].O[N+]([O-])=O.O[N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10724751
SDS View Documents
Fisher Chemical
C/1882/53
500 g
54.10 €
500g
Please to purchase this item. Need a web account? Register with us today!
10642261
SDS View Documents
Fisher Chemical
C/1882/60
1 kg
99.50 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

13477-34-4
238.16
DKCJOKYHYIAZQH-UHFFFAOYSA-N
O.O.O.O.[Ca++].O[N+]([O-])=O.O[N+]([O-])=O
CaH10N2O10
149604
calcium bis(nitric acid) tetrahydrate

Specifications

132°C
13477-34-4
5 to 7
CaH10N2O10
DKCJOKYHYIAZQH-UHFFFAOYSA-N
calcium bis(nitric acid) tetrahydrate
236.15g/mol
45°C
White
Crystalline Solid
149604
O.O.O.O.[Ca++].O[N+]([O-])=O.O[N+]([O-])=O
238.16
Plastic powder jar
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Calcium nitrate tetrahydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.