Learn More
Calcium nitrate hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
43.80 € - 458.00 €
Chemical Identifiers
CAS | 35054-52-5 |
---|---|
Molecular Formula | CaN2O6 |
Molecular Weight (g/mol) | 164.09 |
MDL Number | MFCD00149604 |
InChI Key | ZCCIPPOKBCJFDN-UHFFFAOYSA-N |
Synonym | calcium nitrate hydrate, calcium hydrate dinitrate, acmc-20alid, calcium nitrate monohydrate, ca.2no3.h2o, nitric acid calcium salt hydrate, calcium nitrate-water 1/2/1, nitric acid, calciumsalt, hydrate 9ci |
PubChem CID | 16211464 |
SMILES | [Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11340929
|
Thermo Scientific Alfa Aesar
010683.06 |
5 g | |||||||
11320929
|
Thermo Scientific Alfa Aesar
010683.14 |
25 g |
152.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11330929
|
Thermo Scientific Alfa Aesar
010683.22 |
100 g |
458.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium nitrate hydrate is used as a nitrate enriching salt. It is also used as a component in fertilizers, waste water treatment to prevent odor and inset accelerating concrete admixtures. Further, it is used for regenerable cold packs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
35054-52-5 | |
164.09 | |
ZCCIPPOKBCJFDN-UHFFFAOYSA-N | |
16211464 |
CaN2O6 | |
MFCD00149604 | |
calcium nitrate hydrate, calcium hydrate dinitrate, acmc-20alid, calcium nitrate monohydrate, ca.2no3.h2o, nitric acid calcium salt hydrate, calcium nitrate-water 1/2/1, nitric acid, calciumsalt, hydrate 9ci | |
[Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
132°C (decomposition) | |
35054-52-5 | |
CaN2O6 | |
UN1454 | |
calcium nitrate hydrate, calcium hydrate dinitrate, acmc-20alid, calcium nitrate monohydrate, ca.2no3.h2o, nitric acid calcium salt hydrate, calcium nitrate-water 1/2/1, nitric acid, calciumsalt, hydrate 9ci | |
[Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
164.09 | |
164.09 (Anhydrous) | |
Puratronic™ | |
1.82g/mL | |
Calcium nitrate hydrate |
42°C | |
Crystalline Aggregates | |
MFCD00149604 | |
14,1682 | |
ZCCIPPOKBCJFDN-UHFFFAOYSA-N | |
calcium;dinitrate;hydrate | |
16211464 | |
99.995% | |
Hygroscopic | |
(metals basis) |
Safety and Handling
GHS H Statement
H272-H302-H315-H319-H335
May intensify fire
oxidizer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CALCIUM NITRATE
missing translation for 'rtecsNumber' : EW3000000
missing translation for 'tsca' : TSCA
RUO – Research Use Only