missing translation for 'onlineSavingsMsg'
Learn More

Calcium molybdenum oxide, Puratronic™, 99.9965% (metals basis), Thermo Scientific™

Product Code. 11300929
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-82-4
Molecular Formula CaMoO4
Molecular Weight (g/mol) 200.02
MDL Number MFCD00015976
InChI Key BIOOACNPATUQFW-UHFFFAOYSA-N
Synonym calcium molybdate, molybdate, calcium, calcium molybdate vi, unii-q03c83v2an, calcium molybdenum oxide, molybdic acid h2moo4 , calcium salt 1:1, molybdate moo42-, calcium 1:1 , t-4, molybdate moo42-, calcium 1:1 , beta-4, calcium dioxido dioxo molybdenum, calcium 2+ molybdate
PubChem CID 24620
IUPAC Name calcium;dioxido(dioxo)molybdenum
SMILES [Ca++].[O-][Mo]([O-])(=O)=O

Specifications

Melting Point 965°C
Physical Form Powder
Quantity 25 g
Merck Index 14,1681
Formula Weight 200.01
Percent Purity 99.9965%
Grade Puratronic™
Odor Odorless
Density 4.38 g/mL to 4.53 g/mL
Assay (metals basis)
Chemical Name or Material Calcium molybdenum oxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'