missing translation for 'onlineSavingsMsg'
Learn More

Calcium iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

210.05 € - 636.17 €

Chemical Identifiers

CAS 10102-68-8
Molecular Formula CaI2
Molecular Weight (g/mol) 293.887
MDL Number MFCD00010910
InChI Key UNMYWSMUMWPJLR-UHFFFAOYSA-L
Synonym calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g
PubChem CID 5462722
ChEBI CHEBI:81718
IUPAC Name calcium;diiodide
SMILES [Ca+2].[I-].[I-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11359788
View Documents
Thermo Scientific Alfa Aesar
044254.14
25 g
210.05 €
25g
Please to purchase this item. Need a web account? Register with us today!
11369788
View Documents
Thermo Scientific Alfa Aesar
044254.22
100 g
636.17 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used in chemical research and as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

10102-68-8
293.887
UNMYWSMUMWPJLR-UHFFFAOYSA-L
5462722
calcium;diiodide
CaI2
MFCD00010910
calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g
CHEBI:81718
[Ca+2].[I-].[I-]

Specifications

1100°C
10102-68-8
Beads
MFCD00010910
calcium iodide, calcium diiodide, calciumiodide, calcium 2+ diiodide, acmc-1c61l, ksc491a2n, calcium iodide, anhydrous 25g
UNMYWSMUMWPJLR-UHFFFAOYSA-L
calcium;diiodide
5462722
293.89
Odorless
Hygroscopic; Light sensitive
(metals basis)
784°C
White to Yellow
CaI2
14,1677
Soluble in water,ethyl alcohol,methyl alcohol and acetone. Insoluble in ethyl ether.
[Ca+2].[I-].[I-]
293.887
CHEBI:81718
99.999%
Ampule
3.956 g/mL
Calcium iodide, Ultra dry
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'einecsNumber' : 233-276-8

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.