Learn More
Calcium hypochlorite, tech., granular, 65% available chlorine, Thermo Scientific Chemicals
20.40 € - 68.04 €
Chemical Identifiers
CAS | 7778-54-3 |
---|---|
Molecular Formula | CaCl2O2 |
Molecular Weight (g/mol) | 142.98 |
MDL Number | MFCD00010900 |
InChI Key | ZKQDCIXGCQPQNV-UHFFFAOYSA-N |
Synonym | calcium hypochlorite, calcium hypochloride, bleaching powder, chlorinated lime, hypochlorous acid, calcium salt, caporit, losantin, pittchlor, pittcide, pittclor |
PubChem CID | 24504 |
IUPAC Name | calcium;dihypochlorite |
SMILES | [O-]Cl.[O-]Cl.[Ca+2] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10195310
|
Thermo Scientific Acros
300341000 |
100 g | Plastic bottle |
20.40 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10626291
|
Thermo Scientific Acros
300340010 |
1 kg | Plastic bottle |
68.04 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
7778-54-3 | |
142.98 | |
ZKQDCIXGCQPQNV-UHFFFAOYSA-N | |
24504 | |
[O-]Cl.[O-]Cl.[Ca+2] |
CaCl2O2 | |
MFCD00010900 | |
calcium hypochlorite, calcium hypochloride, bleaching powder, chlorinated lime, hypochlorous acid, calcium salt, caporit, losantin, pittchlor, pittcide, pittclor | |
calcium;dihypochlorite |
Specifications
100°C | |
Green or White | |
Granular Solid or Granules | |
Ca(OCl)2 | |
11,107 | |
calcium hypochlorite, calcium hypochloride, bleaching powder, chlorinated lime, hypochlorous acid, calcium salt, caporit, losantin, pittchlor, pittcide, pittclor | |
ZKQDCIXGCQPQNV-UHFFFAOYSA-N | |
calcium;dihypochlorite | |
24504 | |
≥65% (available Cl) | |
Calcium hypochlorite, tech. |
7778-54-3 | |
∼11.6 (1% aq. soln.) | |
CaCl2O2 | |
MFCD00010900 | |
15,1676 | |
Solubility in water: 200g/L (20°C) (decomposes). Other solubilities: decomposes in alcohol | |
[O-]Cl.[O-]Cl.[Ca+2] | |
142.98 | |
142.98 | |
Technical |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
May intensify fire; oxidizer.
Contact with acids liberates toxic gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-908-7
missing translation for 'rtecsNumber' : NH3485000
missing translation for 'tsca' : TSCA
- Calcium hypochlorite
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
- H272-May intensify fire; oxidiser.
- EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.