missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, Reag. Ph. Eur., puriss. p.a., Honeywell Fluka™

Product Code. 15633300
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
6 x 1 kg

Puriss. p.a., Reag. Ph. Eur.,≥96%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
Synonym calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
PubChem CID 6093208
ChEBI CHEBI:31341
IUPAC Name calcium;dihydroxide
SMILES [OH-].[OH-].[Ca++]

Specifications

Quantity 25 kg
Linear Formula Ca(OH)2
UN Number NONH for all modes of transport
Formula Weight 74.09g/mol
Percent Purity ≥96%
Grade puriss. p.a.
Density 2.24g/mL at 25°C
Chemical Name or Material Calcium Hydroxide, Reag. Ph. Eur.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.