missing translation for 'onlineSavingsMsg'
Learn More

Calcium Hydroxide, Extra Pure, SLR, Fisher Chemical™

94.50 € - 257.00 €

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
IUPAC Name calcium dihydroxide
SMILES [OH-].[OH-].[Ca++]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10293320
SDS View Documents
Fisher Chemical
C/1640/60
1 kg Plastic powder jar N/A N/A N/A
10735901
SDS View Documents
Fisher Chemical
C/1640/65
5 kg Plastic keg N/A N/A N/A
Description

Description

Specifications

Chemical Identifiers

1305-62-0
74.09
AXCZMVOFGPJBDE-UHFFFAOYSA-L
[OH-].[OH-].[Ca++]
CaH2O2
MFCD00010901
calcium dihydroxide

Specifications

2850°C
1305-62-0
12.4
CaH2O2
AXCZMVOFGPJBDE-UHFFFAOYSA-L
calcium dihydroxide
74.09g/mol
580°C
White
Solid
MFCD00010901
[OH-].[OH-].[Ca++]
74.09
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium Hydroxide, Extra Pure, SLR, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.