missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydrogen phosphate, anhydrous, 98% min, Thermo Scientific Chemicals

66.93 € - 428.13 €

Chemical Identifiers

CAS 7757-93-9
Molecular Formula CaHO4P
Molecular Weight (g/mol) 136.06
MDL Number MFCD00010909
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
Synonym calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1
PubChem CID 24441
ChEBI CHEBI:32596
IUPAC Name calcium;hydrogen phosphate
SMILES [Ca++].OP([O-])([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11399387
View Documents
Thermo Scientific Alfa Aesar
040232.30
250 g
66.93 €
250g
Please to purchase this item. Need a web account? Register with us today!
11309397
View Documents
Thermo Scientific Alfa Aesar
040232.A1
1 kg
132.66 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11389387
View Documents
Thermo Scientific Alfa Aesar
040232.A7
5 kg
428.13 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as a feed additive for livestock and poultry. It can promote feed digestion, put on animal’s weight, raise laying rate or producing milk rate, cure diseases such as rickets, osteomalacia and anemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7757-93-9
136.06
FUFJGUQYACFECW-UHFFFAOYSA-L
24441
calcium;hydrogen phosphate
CaHO4P
MFCD00010909
calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1
CHEBI:32596
[Ca++].OP([O-])([O-])=O

Specifications

7757-93-9
Powder
CaHPO4
14,1692
Soluble in hydrochloric acid,nitric acid,acetic acid,ammonium citrate solution. Slightly soluble in water. Insoluble in ethanol.
[Ca++].OP([O-])([O-])=O
136.06
CHEBI:32596
≤98%
Calcium hydrogen phosphate, Anhydrous
White
CaHO4P
MFCD00010909
calcium hydrogen phosphate, calcium hydrogenphosphate, calcium phosphate dibasic, calcium phosphate, dibasic, biofos, fujicalin s, dicafos an, dibasic calcium phosphate, monocalcium acid phosphate, phosphoric acid, calcium salt 1:1
FUFJGUQYACFECW-UHFFFAOYSA-L
calcium;hydrogen phosphate
24441
136.06
Odorless
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-826-1

missing translation for 'rtecsNumber' : TB8528000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.