missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Hardness Indicator, No Murexide, 100 Tablets, Fisher Chemical
Click to view available options
100tablets, Glass Jar
Specifications
Specifications
Color | Blue-Purple |
Physical Form | Solid |
Quantity | 100 Tablets |
MDL Number | 12777 |
Packaging | Glass Jar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Calcium Hardness Indicator, No Murexide, 100 Tablets, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction