missing translation for 'onlineSavingsMsg'
Learn More

Calcium glycerophosphate hydrate, 97%, Thermo Scientific Chemicals

Product Code. 11437737
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28917-82-0
Molecular Formula C3H7CaO6P
Molecular Weight (g/mol) 210.14
MDL Number MFCD00042643
InChI Key IWIRHXNCFWGFJE-UHFFFAOYNA-L
Synonym calcium glycerophosphate hydrate, calcii glycerophosphas, c3h7o6p.ca.h2o, glycerophosphoric acid calcium salt hydrate, calcium 2,3-dihydroxypropyl phosphate hydrate, calcium dl-glycerol 3-phosphate hydrate
PubChem CID 71311563
IUPAC Name calcium 3-(phosphonatooxy)propane-1,2-diol
SMILES [Ca++].OCC(O)COP([O-])([O-])=O

Specifications

Quantity 250 g
Merck Index 14,1670
Formula Weight 210.15 (Anhydrous)
Percent Purity 97%
Chemical Name or Material Calcium glycerophosphate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.