missing translation for 'onlineSavingsMsg'
Learn More

Calcium glycerophosphate hydrate, 97%, Thermo Scientific Chemicals

Product Code. 11427737
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28917-82-0
Molecular Formula C3H7CaO6P
Molecular Weight (g/mol) 210.14
MDL Number MFCD00042643
InChI Key IWIRHXNCFWGFJE-UHFFFAOYNA-L
Synonym calcium glycerophosphate hydrate, calcii glycerophosphas, c3h7o6p.ca.h2o, glycerophosphoric acid calcium salt hydrate, calcium 2,3-dihydroxypropyl phosphate hydrate, calcium dl-glycerol 3-phosphate hydrate
PubChem CID 71311563
IUPAC Name calcium 3-(phosphonatooxy)propane-1,2-diol
SMILES [Ca++].OCC(O)COP([O-])([O-])=O

Specifications

Quantity 1000 g
Merck Index 14,1670
Formula Weight 210.15 (Anhydrous)
Percent Purity 97%
Chemical Name or Material Calcium glycerophosphate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'