missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Chloride Dihydrate, ACS Reagent, Honeywell Fluka™
ACS Reagent,≥99%
32.67 € - 1345.00 €
Chemical Identifiers
CAS | 10035-04-8 |
---|---|
Molecular Formula | CaCl2 · 2H2O |
MDL Number | MFCD00149613 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15672220
|
Honeywell-Fluka
223506-25G |
25 g | |||||||
15692220
|
Honeywell-Fluka
223506-500G |
500 g | |||||||
15662220
|
Honeywell-Fluka
223506-2.5KG |
2.5 kg | |||||||
15602230
|
Honeywell-Fluka
223506-6X500G |
6 x 500 g | |||||||
15682220
|
Honeywell-Fluka
223506-4X2.5KG |
4 x 2.5 kg | |||||||
15652220
|
Honeywell-Fluka
223506-12KG |
12 kg | |||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
10035-04-8 | |
CaCl2 · 2H2O | |
MFCD00149613 |
Specifications
10035-04-8 | |
MFCD00149613 | |
147.01g/mol | |
ACS Reagent | |
Calcium Chloride Dihydrate |
CaCl2 · 2H2O | |
NONH for all modes of transport | |
≥99% | |
0.01mmHg (20°C) |
Safety and Handling
P305 + P351 + P338
H319
missing translation for 'einecsNumber' : 233-140-8
missing translation for 'rtecsNumber' : EV9810000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Calcium chloride dihydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.