missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride Dihydrate, ≥99.0%, ReagentPlus™ Grade, Honeywell™

Product Code. 15667750
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
12 kg
Packaging:
Drum with liner
Fiberboard box
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-04-8
MDL Number MFCD00149613

Specifications

Boiling Point 1.90°C (1.013hPa)
Melting Point 765°C
Color Colorless
pH 4,5 - 8
Physical Form Solid
Quantity 1 kg
UN Number NONH for all modes of transport
Formula Weight 147.01g/mol
Percent Purity ≥99.0%
Odor Odorless
Packaging Poly bottle
Chemical Name or Material Calcium chloride dihydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'